Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604810 MIM: 604812 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LILRA1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
LILRA1 - leukocyte immunoglobulin like receptor A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LILRA2 - leukocyte immunoglobulin like receptor A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130917.2 | Intron | NP_001124389.2 | ||||
NM_001290270.1 | Intron | NP_001277199.1 | ||||
NM_001290271.1 | Intron | NP_001277200.1 | ||||
NM_006866.3 | Intron | NP_006857.2 | ||||
XM_006722986.1 | Intron | XP_006723049.1 | ||||
XM_011526385.1 | Intron | XP_011524687.1 | ||||
XM_011526386.1 | Intron | XP_011524688.1 | ||||
XM_011526387.1 | Intron | XP_011524689.1 | ||||
XM_011526388.1 | Intron | XP_011524690.1 | ||||
XM_011526389.1 | Intron | XP_011524691.1 | ||||
XM_011526390.1 | Intron | XP_011524692.1 | ||||
XM_011526391.1 | Intron | XP_011524693.1 | ||||
XM_017026224.1 | Intron | XP_016881713.1 |
Set Membership: |
HapMap |