Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604812 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LILRA2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LILRA2 - leukocyte immunoglobulin like receptor A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130917.2 | 743 | Silent Mutation | CTC,CTT | L,L 53 | NP_001124389.2 | |
NM_001290270.1 | 743 | Silent Mutation | CTC,CTT | L,L 41 | NP_001277199.1 | |
NM_001290271.1 | 743 | Silent Mutation | CTC,CTT | L,L 53 | NP_001277200.1 | |
NM_006866.3 | 743 | Silent Mutation | CTC,CTT | L,L 53 | NP_006857.2 | |
XM_006722986.1 | 743 | Silent Mutation | CTC,CTT | L,L 53 | XP_006723049.1 | |
XM_011526385.1 | 743 | Silent Mutation | CTC,CTT | L,L 53 | XP_011524687.1 | |
XM_011526386.1 | 743 | Silent Mutation | CTC,CTT | L,L 41 | XP_011524688.1 | |
XM_011526387.1 | 743 | Silent Mutation | CTC,CTT | L,L 41 | XP_011524689.1 | |
XM_011526388.1 | 743 | Silent Mutation | CTC,CTT | L,L 41 | XP_011524690.1 | |
XM_011526389.1 | 743 | Intron | XP_011524691.1 | |||
XM_011526390.1 | 743 | Silent Mutation | CTC,CTT | L,L 53 | XP_011524692.1 | |
XM_011526391.1 | 743 | Silent Mutation | CTC,CTT | L,L 53 | XP_011524693.1 | |
XM_017026224.1 | 743 | Silent Mutation | CTC,CTT | L,L 53 | XP_016881713.1 |
Set Membership: |
HapMap |