Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616838 MIM: 603842 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC17A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CLEC17A - C-type lectin domain family 17 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204118.1 | 68 | Intron | NP_001191047.1 | |||
NM_207390.3 | 68 | Intron | NP_997273.3 | |||
XM_017026785.1 | 68 | Intron | XP_016882274.1 | |||
XM_017026786.1 | 68 | Intron | XP_016882275.1 | |||
XM_017026787.1 | 68 | Intron | XP_016882276.1 | |||
XM_017026788.1 | 68 | UTR 5 | XP_016882277.1 | |||
XM_017026789.1 | 68 | Intron | XP_016882278.1 | |||
XM_017026790.1 | 68 | Intron | XP_016882279.1 | |||
XM_017026791.1 | 68 | Intron | XP_016882280.1 | |||
XM_017026792.1 | 68 | Intron | XP_016882281.1 | |||
XM_017026793.1 | 68 | Intron | XP_016882282.1 | |||
XM_017026794.1 | 68 | Intron | XP_016882283.1 |
NDUFB7 - NADH:ubiquinone oxidoreductase subunit B7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |