Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300579 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHROOM4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SHROOM4 - shroom family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020717.3 | 8147 | UTR 3 | NP_065768.2 | |||
XM_017029682.1 | 8147 | Intron | XP_016885171.1 | |||
XM_017029683.1 | 8147 | Intron | XP_016885172.1 | |||
XM_017029684.1 | 8147 | Intron | XP_016885173.1 | |||
XM_017029685.1 | 8147 | Intron | XP_016885174.1 | |||
XM_017029686.1 | 8147 | Intron | XP_016885175.1 | |||
XM_017029687.1 | 8147 | Intron | XP_016885176.1 |
Set Membership: |
HapMap |