Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDKL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDKL4 - cyclin dependent kinase like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009565.1 | Intron | NP_001009565.1 | ||||
XM_011532815.2 | Intron | XP_011531117.1 | ||||
XM_011532816.2 | Intron | XP_011531118.1 | ||||
XM_011532817.2 | Intron | XP_011531119.1 | ||||
XM_011532819.2 | Intron | XP_011531121.1 | ||||
XM_017003981.1 | Intron | XP_016859470.1 | ||||
XM_017003982.1 | Intron | XP_016859471.1 | ||||
XM_017003983.1 | Intron | XP_016859472.1 | ||||
XM_017003984.1 | Intron | XP_016859473.1 | ||||
XM_017003985.1 | Intron | XP_016859474.1 |
Set Membership: |
HapMap |