Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147558 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGB6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ITGB6 - integrin subunit beta 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000888.4 | 2466 | UTR 3 | NP_000879.2 | |||
NM_001282353.1 | 2466 | UTR 3 | NP_001269282.1 | |||
NM_001282354.1 | 2466 | UTR 3 | NP_001269283.1 | |||
NM_001282355.1 | 2466 | UTR 3 | NP_001269284.1 | |||
NM_001282388.1 | 2466 | UTR 3 | NP_001269317.1 | |||
NM_001282389.1 | 2466 | UTR 3 | NP_001269318.1 | |||
NM_001282390.1 | 2466 | UTR 3 | NP_001269319.1 |
Set Membership: |
HapMap |