Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGGAAGTCCTCAAATTCAAAGCA[A/G]GCTGATTTCCAAGCATTCCAAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602310 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100505984 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100505984 - uncharacterized LOC100505984 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBMS1 - RNA binding motif single stranded interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002897.4 | Intron | NP_002888.1 | ||||
NM_016836.3 | Intron | NP_058520.1 | ||||
XM_005246737.1 | Intron | XP_005246794.1 | ||||
XM_005246738.2 | Intron | XP_005246795.1 | ||||
XM_005246739.2 | Intron | XP_005246796.1 | ||||
XM_005246740.1 | Intron | XP_005246797.1 | ||||
XM_005246741.3 | Intron | XP_005246798.1 | ||||
XM_017004626.1 | Intron | XP_016860115.1 |