Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACGAAGAACAGGATCCAGGCCA[A/G]GTAGACAGAACCAGCGAGAGACACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610561 MIM: 608547 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRSS53 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
PRSS53 - protease, serine 53 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VKORC1 - vitamin K epoxide reductase complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001311311.1 | 712 | Silent Mutation | CTG,TTG | L,L 148 | NP_001298240.1 | |
NM_024006.5 | 712 | Silent Mutation | CTG,TTG | L,L 120 | NP_076869.1 | |
NM_206824.2 | 712 | Missense Mutation | CCT,CTT | P,L 83 | NP_996560.1 |
ZNF646 - zinc finger protein 646 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap DME Validated Inventoried |