Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
49 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 150330 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMNA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LMNA - lamin A/C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257374.2 | Intron | NP_001244303.1 | ||||
NM_001282624.1 | Intron | NP_001269553.1 | ||||
NM_001282625.1 | Intron | NP_001269554.1 | ||||
NM_001282626.1 | Intron | NP_001269555.1 | ||||
NM_005572.3 | Intron | NP_005563.1 | ||||
NM_170707.3 | Intron | NP_733821.1 | ||||
NM_170708.3 | Intron | NP_733822.1 | ||||
XM_011509533.1 | Intron | XP_011507835.1 | ||||
XM_011509534.1 | Intron | XP_011507836.1 |