Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 113995 MIM: 609949 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5AR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C5AR1 - complement component 5a receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C5AR2 - complement component 5a receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271749.1 | 376 | Intron | NP_001258678.1 | |||
NM_001271750.1 | 376 | Intron | NP_001258679.1 | |||
NM_018485.2 | 376 | Intron | NP_060955.1 | |||
XM_011526736.2 | 376 | UTR 5 | XP_011525038.1 |
Set Membership: |
HapMap |