Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTATTTTTTCCTTTGTGATAGG[C/T]AGGAAACTGAATTTTTTTTTAAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602680 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARHGAP5 - Rho GTPase activating protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030055.1 | Intron | NP_001025226.1 | ||||
NM_001173.2 | Intron | NP_001164.2 | ||||
XM_005267635.3 | Intron | XP_005267692.1 | ||||
XM_005267636.3 | Intron | XP_005267693.1 | ||||
XM_017021287.1 | Intron | XP_016876776.1 | ||||
XM_017021288.1 | Intron | XP_016876777.1 |
ARHGAP5-AS1 - ARHGAP5 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |