Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180465 MIM: 602671 MIM: 610971 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC84 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC84 - coiled-coil domain containing 84 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198489.2 | Intron | NP_940891.1 | ||||
XM_011542799.1 | Intron | XP_011541101.1 | ||||
XM_011542800.2 | Intron | XP_011541102.1 | ||||
XM_017017652.1 | Intron | XP_016873141.1 |
MIR3656 - microRNA 3656 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS25 - ribosomal protein S25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001028.2 | Intron | NP_001019.1 |
SLC37A4 - solute carrier family 37 member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAPPC4 - trafficking protein particle complex 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |