Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179616 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf74 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C11orf74 - chromosome 11 open reading frame 74 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276722.1 | Intron | NP_001263651.1 | ||||
NM_001276723.1 | Intron | NP_001263652.1 | ||||
NM_001276724.1 | Intron | NP_001263653.1 | ||||
NM_001276725.1 | Intron | NP_001263654.1 | ||||
NM_001276726.1 | Intron | NP_001263655.1 | ||||
NM_001276727.1 | Intron | NP_001263656.1 | ||||
NM_138787.3 | Intron | NP_620142.2 | ||||
XM_006718138.3 | Intron | XP_006718201.1 | ||||
XM_011519885.2 | Intron | XP_011518187.1 |
RAG2 - recombination activating gene 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000536.3 | Intron | NP_000527.2 | ||||
NM_001243785.1 | Intron | NP_001230714.1 | ||||
NM_001243786.1 | Intron | NP_001230715.1 |