Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601142 MIM: 607215 | ||||||||||||||||||||
Literature Links: |
KCNAB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNAB2 - potassium voltage-gated channel subfamily A regulatory beta subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199860.1 | Intron | NP_001186789.1 | ||||
NM_001199861.1 | Intron | NP_001186790.1 | ||||
NM_001199862.1 | Intron | NP_001186791.1 | ||||
NM_001199863.1 | Intron | NP_001186792.1 | ||||
NM_003636.3 | Intron | NP_003627.1 | ||||
NM_172130.2 | Intron | NP_742128.1 | ||||
XM_005263514.2 | Intron | XP_005263571.1 | ||||
XM_011542321.2 | Intron | XP_011540623.1 | ||||
XM_011542322.2 | Intron | XP_011540624.1 | ||||
XM_017002618.1 | Intron | XP_016858107.1 | ||||
XM_017002619.1 | Intron | XP_016858108.1 | ||||
XM_017002620.1 | Intron | XP_016858109.1 | ||||
XM_017002621.1 | Intron | XP_016858110.1 |
NPHP4 - nephronophthisis 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |