Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607365 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPH PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LIPH - lipase H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139248.2 | 1777 | UTR 3 | NP_640341.1 | |||
XM_006713529.3 | 1777 | UTR 3 | XP_006713592.1 | |||
XM_011512530.2 | 1777 | UTR 3 | XP_011510832.1 | |||
XM_011512531.2 | 1777 | UTR 3 | XP_011510833.1 | |||
XM_017005852.1 | 1777 | UTR 3 | XP_016861341.1 |
TMEM41A - transmembrane protein 41A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |