Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616613 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL6A6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
COL6A6 - collagen type VI alpha 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102608.1 | 5975 | Intron | NP_001096078.1 | |||
XM_005247121.4 | 5975 | Intron | XP_005247178.1 | |||
XM_011512423.2 | 5975 | Intron | XP_011510725.1 | |||
XM_011512424.2 | 5975 | Intron | XP_011510726.1 | |||
XM_011512425.2 | 5975 | Intron | XP_011510727.1 | |||
XM_011512426.2 | 5975 | Intron | XP_011510728.1 | |||
XM_011512428.2 | 5975 | Intron | XP_011510730.1 | |||
XM_011512431.2 | 5975 | Intron | XP_011510733.1 | |||
XM_011512432.2 | 5975 | Intron | XP_011510734.1 | |||
XM_017005711.1 | 5975 | Intron | XP_016861200.1 | |||
XM_017005712.1 | 5975 | Intron | XP_016861201.1 | |||
XM_017005713.1 | 5975 | Intron | XP_016861202.1 | |||
XM_017005714.1 | 5975 | UTR 5 | XP_016861203.1 | |||
XM_017005715.1 | 5975 | Intron | XP_016861204.1 | |||
XM_017005716.1 | 5975 | Intron | XP_016861205.1 |
Set Membership: |
HapMap |