Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194553 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF80 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF80 - zinc finger protein 80 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007136.3 | 3331 | Intron | NP_009067.2 | |||
XM_017007132.1 | 3331 | Intron | XP_016862621.1 | |||
XM_017007133.1 | 3331 | UTR 3 | XP_016862622.1 | |||
XM_017007134.1 | 3331 | UTR 3 | XP_016862623.1 | |||
XM_017007135.1 | 3331 | Intron | XP_016862624.1 | |||
XM_017007136.1 | 3331 | Intron | XP_016862625.1 |
Set Membership: |
HapMap |