Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601645 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP2R5C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP2R5C - protein phosphatase 2 regulatory subunit B'gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161725.1 | Intron | NP_001155197.1 | ||||
NM_001161726.1 | Intron | NP_001155198.1 | ||||
NM_002719.3 | Intron | NP_002710.2 | ||||
NM_178586.2 | Intron | NP_848701.1 | ||||
NM_178587.2 | Intron | NP_848702.1 | ||||
XM_005267819.1 | Intron | XP_005267876.1 | ||||
XM_005267820.1 | Intron | XP_005267877.1 | ||||
XM_005267822.3 | Intron | XP_005267879.1 | ||||
XM_005267823.1 | Intron | XP_005267880.1 | ||||
XM_005267824.1 | Intron | XP_005267881.1 | ||||
XM_005267826.1 | Intron | XP_005267883.1 | ||||
XM_005267827.1 | Intron | XP_005267884.1 | ||||
XM_011536914.2 | Intron | XP_011535216.1 | ||||
XM_011536915.2 | Intron | XP_011535217.1 | ||||
XM_011536916.2 | Intron | XP_011535218.1 | ||||
XM_011536918.1 | Intron | XP_011535220.1 | ||||
XM_011536919.2 | Intron | XP_011535221.1 | ||||
XM_011536920.1 | Intron | XP_011535222.1 | ||||
XM_011536921.2 | Intron | XP_011535223.1 | ||||
XM_017021422.1 | Intron | XP_016876911.1 | ||||
XM_017021423.1 | Intron | XP_016876912.1 | ||||
XM_017021424.1 | Intron | XP_016876913.1 |