Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608834 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CREB3L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CREB3L2 - cAMP responsive element binding protein 3 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253775.1 | 4161 | Intron | NP_001240704.1 | |||
NM_001318246.1 | 4161 | UTR 3 | NP_001305175.1 | |||
NM_194071.3 | 4161 | UTR 3 | NP_919047.2 | |||
XM_005250541.4 | 4161 | UTR 3 | XP_005250598.1 | |||
XM_017012540.1 | 4161 | UTR 3 | XP_016868029.1 | |||
XM_017012541.1 | 4161 | UTR 3 | XP_016868030.1 |