Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCTGGCCTCGTAGAGCGAGCGGC[A/G]GCCCCGATGCGTCCTCTCATCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600197 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100128653 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100128653 - uncharacterized LOC100128653 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAFK - MAF bZIP transcription factor K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002360.3 | Intron | NP_002351.1 | ||||
XM_005249851.2 | Intron | XP_005249908.2 | ||||
XM_006715773.2 | Intron | XP_006715836.1 |
TMEM184A - transmembrane protein 184A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |