Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600312 MIM: 614905 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6836 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR6836 - microRNA 6836 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUDT1 - nudix hydrolase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNX8 - sorting nexin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013321.3 | 1673 | UTR 3 | NP_037453.1 | |||
XM_011515329.2 | 1673 | UTR 3 | XP_011513631.1 | |||
XM_011515330.1 | 1673 | UTR 3 | XP_011513632.1 | |||
XM_017012083.1 | 1673 | UTR 3 | XP_016867572.1 | |||
XM_017012084.1 | 1673 | UTR 3 | XP_016867573.1 |
Set Membership: |
HapMap |