Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614506 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRAT1 - BRCA1 associated ATM activator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IQCE - IQ motif containing E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287499.1 | 152 | UTR 5 | NP_001274428.1 | |||
NM_001287500.1 | 152 | UTR 5 | NP_001274429.1 | |||
NM_001287501.1 | 152 | UTR 5 | NP_001274430.1 | |||
NM_001287502.1 | 152 | UTR 5 | NP_001274431.1 | |||
NM_152558.4 | 152 | UTR 5 | NP_689771.3 | |||
XM_006715676.2 | 152 | Intron | XP_006715739.1 | |||
XM_011515242.1 | 152 | UTR 5 | XP_011513544.1 | |||
XM_011515243.1 | 152 | Intron | XP_011513545.1 | |||
XM_011515244.2 | 152 | Intron | XP_011513546.1 | |||
XM_017011902.1 | 152 | UTR 5 | XP_016867391.1 | |||
XM_017011903.1 | 152 | Intron | XP_016867392.1 |