Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600935 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNJ8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KCNJ8 - potassium voltage-gated channel subfamily J member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004982.3 | 2221 | UTR 3 | NP_004973.1 | |||
XM_005253358.4 | 2221 | UTR 3 | XP_005253415.1 | |||
XM_017019283.1 | 2221 | UTR 3 | XP_016874772.1 | |||
XM_017019284.1 | 2221 | UTR 3 | XP_016874773.1 |
Set Membership: |
HapMap |