Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610229 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPN14 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CAPN14 - calpain 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145122.1 | 2304 | UTR 3 | NP_001138594.1 | |||
NM_001321270.1 | 2304 | UTR 3 | NP_001308199.1 | |||
XM_011532863.2 | 2304 | UTR 3 | XP_011531165.1 | |||
XM_011532864.2 | 2304 | UTR 3 | XP_011531166.1 | |||
XM_011532865.1 | 2304 | UTR 3 | XP_011531167.1 | |||
XM_011532866.2 | 2304 | UTR 3 | XP_011531168.1 |
Set Membership: |
HapMap |