Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608225 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT14 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GALNT14 - polypeptide N-acetylgalactosaminyltransferase 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253826.1 | Intron | NP_001240755.1 | ||||
NM_001253827.1 | Intron | NP_001240756.1 | ||||
NM_024572.3 | Intron | NP_078848.2 | ||||
XM_011533105.1 | Intron | XP_011531407.1 | ||||
XM_017004906.1 | Intron | XP_016860395.1 | ||||
XM_017004907.1 | Intron | XP_016860396.1 | ||||
XM_017004908.1 | Intron | XP_016860397.1 |
Set Membership: |
HapMap |