Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAACAATTTGTTAACACCAGTAA[C/G]AATTTGCTGTTGCTGTTTAAAACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171500 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACP1 - acid phosphatase 1, soluble | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM150B - family with sequence similarity 150 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002919.2 | Intron | NP_001002919.2 | ||||
XM_005264689.4 | Intron | XP_005264746.1 | ||||
XM_006711879.3 | Intron | XP_006711942.1 | ||||
XM_011510342.2 | Intron | XP_011508644.1 |
LOC101927262 - melanoma-associated antigen C1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |