Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTACACGTGTGCACAAAGAGAT[A/T]TATACCAAGAGGTTTACTATCGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114080 MIM: 607049 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CAMK4 - calcium/calmodulin dependent protein kinase IV | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STARD4 - StAR related lipid transfer domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308056.1 | Intron | NP_001294985.1 | ||||
NM_001308057.1 | Intron | NP_001294986.1 | ||||
NM_001308058.1 | Intron | NP_001294987.1 | ||||
NM_001308059.1 | Intron | NP_001294988.1 | ||||
NM_001308060.1 | Intron | NP_001294989.1 | ||||
NM_001308061.1 | Intron | NP_001294990.1 | ||||
NM_139164.2 | Intron | NP_631903.1 | ||||
XM_017009041.1 | Intron | XP_016864530.1 | ||||
XM_017009042.1 | Intron | XP_016864531.1 | ||||
XM_017009043.1 | Intron | XP_016864532.1 | ||||
XM_017009044.1 | Intron | XP_016864533.1 |
STARD4-AS1 - STARD4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |