Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120328 MIM: 600424 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL18A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COL18A1 - collagen type XVIII alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030582.3 | 2902 | Intron | NP_085059.2 | |||
NM_130444.2 | 2902 | Intron | NP_569711.2 | |||
NM_130445.3 | 2902 | Intron | NP_569712.2 |
SLC19A1 - solute carrier family 19 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205206.1 | 2902 | Intron | NP_001192135.1 | |||
NM_001205207.1 | 2902 | Intron | NP_001192136.1 | |||
NM_194255.2 | 2902 | Intron | NP_919231.1 | |||
XM_005261164.3 | 2902 | Intron | XP_005261221.1 | |||
XM_011529696.2 | 2902 | Intron | XP_011527998.1 | |||
XM_011529698.2 | 2902 | Intron | XP_011528000.1 | |||
XM_011529700.2 | 2902 | Intron | XP_011528002.1 | |||
XM_011529701.2 | 2902 | Intron | XP_011528003.1 | |||
XM_011529702.2 | 2902 | Intron | XP_011528004.1 | |||
XM_011529703.2 | 2902 | Intron | XP_011528005.1 | |||
XM_011529704.2 | 2902 | Intron | XP_011528006.1 | |||
XM_011529706.2 | 2902 | Intron | XP_011528008.1 | |||
XM_011529709.2 | 2902 | Intron | XP_011528011.1 | |||
XM_017028443.1 | 2902 | Intron | XP_016883932.1 | |||
XM_017028444.1 | 2902 | Intron | XP_016883933.1 | |||
XM_017028445.1 | 2902 | UTR 3 | XP_016883934.1 | |||
XM_017028446.1 | 2902 | Intron | XP_016883935.1 |
Set Membership: |
HapMap |