Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616374 | ||||||||||||||||||||
Literature Links: |
BEND3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BEND3 - BEN domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080450.2 | 3270 | UTR 3 | NP_001073919.1 | |||
XM_005267079.3 | 3270 | Intron | XP_005267136.1 | |||
XM_005267080.3 | 3270 | UTR 3 | XP_005267137.1 | |||
XM_011536005.2 | 3270 | Intron | XP_011534307.1 | |||
XM_017011138.1 | 3270 | UTR 3 | XP_016866627.1 |