Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601426 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGD5-AS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FGD5-AS1 - FGD5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NR2C2 - nuclear receptor subfamily 2 group C member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291694.1 | Intron | NP_001278623.1 | ||||
NM_003298.4 | Intron | NP_003289.2 | ||||
XM_011534058.2 | Intron | XP_011532360.1 | ||||
XM_011534059.2 | Intron | XP_011532361.1 | ||||
XM_011534061.2 | Intron | XP_011532363.1 | ||||
XM_011534063.2 | Intron | XP_011532365.1 | ||||
XM_011534064.2 | Intron | XP_011532366.1 | ||||
XM_011534065.2 | Intron | XP_011532367.1 | ||||
XM_011534066.2 | Intron | XP_011532368.1 | ||||
XM_017007118.1 | Intron | XP_016862607.1 | ||||
XM_017007119.1 | Intron | XP_016862608.1 | ||||
XM_017007120.1 | Intron | XP_016862609.1 |
Set Membership: |
HapMap |