Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611429 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C3orf35 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C3orf35 - chromosome 3 open reading frame 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302831.1 | Intron | NP_001289760.1 | ||||
NM_001302832.1 | Intron | NP_001289761.1 | ||||
NM_178339.2 | Intron | NP_848029.2 | ||||
NM_178342.2 | Intron | NP_848032.1 | ||||
NM_178344.2 | Intron | NP_848034.2 | ||||
XM_017006291.1 | Intron | XP_016861780.1 | ||||
XM_017006292.1 | Intron | XP_016861781.1 | ||||
XM_017006293.1 | Intron | XP_016861782.1 | ||||
XM_017006294.1 | Intron | XP_016861783.1 | ||||
XM_017006295.1 | Intron | XP_016861784.1 | ||||
XM_017006296.1 | Intron | XP_016861785.1 |