Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611459 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC10A7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC10A7 - solute carrier family 10 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029998.5 | Intron | NP_001025169.1 | ||||
NM_001300842.2 | Intron | NP_001287771.1 | ||||
NM_001317816.1 | Intron | NP_001304745.1 | ||||
NM_001317817.1 | Intron | NP_001304746.1 | ||||
NM_001317818.1 | Intron | NP_001304747.1 | ||||
NM_032128.4 | Intron | NP_115504.1 | ||||
XM_011532311.2 | Intron | XP_011530613.1 | ||||
XM_011532313.2 | Intron | XP_011530615.1 | ||||
XM_011532314.2 | Intron | XP_011530616.1 | ||||
XM_017008689.1 | Intron | XP_016864178.1 | ||||
XM_017008690.1 | Intron | XP_016864179.1 | ||||
XM_017008691.1 | Intron | XP_016864180.1 | ||||
XM_017008692.1 | Intron | XP_016864181.1 |