Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611143 MIM: 606769 MIM: 611983 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM175A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FAM175A - family with sequence similarity 175 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139076.2 | Intron | NP_620775.2 | ||||
XM_005263280.4 | Intron | XP_005263337.1 | ||||
XM_017008694.1 | Intron | XP_016864183.1 |
HELQ - helicase, POLQ-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPS18C - mitochondrial ribosomal protein S18C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |