Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601542 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PITX2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PITX2 - paired like homeodomain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000325.5 | 2859 | UTR 3 | NP_000316.2 | |||
NM_001204397.1 | 2859 | UTR 3 | NP_001191326.1 | |||
NM_001204398.1 | 2859 | UTR 3 | NP_001191327.1 | |||
NM_001204399.1 | 2859 | UTR 3 | NP_001191328.1 | |||
NM_153426.2 | 2859 | UTR 3 | NP_700475.1 | |||
NM_153427.2 | 2859 | UTR 3 | NP_700476.1 | |||
XM_011532027.2 | 2859 | UTR 3 | XP_011530329.1 |
Set Membership: |
HapMap |