Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611798 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFR3A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EFR3A - EFR3 homolog A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323553.1 | Intron | NP_001310482.1 | ||||
NM_001323554.1 | Intron | NP_001310483.1 | ||||
NM_001323555.1 | Intron | NP_001310484.1 | ||||
NM_001323556.1 | Intron | NP_001310485.1 | ||||
NM_001323557.1 | Intron | NP_001310486.1 | ||||
NM_001323558.1 | Intron | NP_001310487.1 | ||||
NM_015137.5 | Intron | NP_055952.2 |