Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603544 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC85C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCDC85C - coiled-coil domain containing 85C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144995.1 | Intron | NP_001138467.1 | ||||
XM_011536705.2 | Intron | XP_011535007.1 | ||||
XM_011536706.2 | Intron | XP_011535008.1 | ||||
XM_011536707.2 | Intron | XP_011535009.1 |
CCNK - cyclin K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |