Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604087 | ||||||||||||||||||||
Literature Links: |
CYP46A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYP46A1 - cytochrome P450 family 46 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006668.1 | Intron | NP_006659.1 | ||||
XM_005267274.4 | Intron | XP_005267331.1 | ||||
XM_011536364.1 | Intron | XP_011534666.1 | ||||
XM_011536365.1 | Intron | XP_011534667.1 | ||||
XM_017020933.1 | Intron | XP_016876422.1 |
HHIPL1 - HHIP like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |