Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZCCHC10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZCCHC10 - zinc finger CCHC-type containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300816.2 | Intron | NP_001287745.1 | ||||
NM_001300817.2 | Intron | NP_001287746.1 | ||||
NM_001300818.2 | Intron | NP_001287747.1 | ||||
NM_001300819.2 | Intron | NP_001287748.1 | ||||
NM_001300822.2 | Intron | NP_001287751.1 | ||||
NM_001308127.1 | Intron | NP_001295056.1 | ||||
NM_001308128.1 | Intron | NP_001295057.1 | ||||
NM_001308129.1 | Intron | NP_001295058.1 | ||||
NM_001308130.1 | Intron | NP_001295059.1 | ||||
NM_017665.3 | Intron | NP_060135.1 | ||||
XM_011543502.1 | Intron | XP_011541804.1 | ||||
XM_011543503.2 | Intron | XP_011541805.1 |