Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610693 MIM: 616283 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HYLS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HYLS1 - HYLS1, centriolar and ciliogenesis associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134793.1 | Intron | NP_001128265.1 | ||||
NM_145014.2 | Intron | NP_659451.1 | ||||
XM_005271430.2 | Intron | XP_005271487.1 | ||||
XM_006718777.3 | Intron | XP_006718840.1 | ||||
XM_011542659.2 | Intron | XP_011540961.1 | ||||
XM_017017320.1 | Intron | XP_016872809.1 | ||||
XM_017017321.1 | Intron | XP_016872810.1 |
PUS3 - pseudouridylate synthase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |