Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATCTAAAATGCAAAATTAGATAT[C/T]GCAATTTAGAATATAAGAGAATTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QTNF7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1QTNF7 - C1q and tumor necrosis factor related protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135170.1 | Intron | NP_001128642.1 | ||||
NM_001135171.1 | Intron | NP_001128643.1 | ||||
NM_031911.4 | Intron | NP_114117.1 | ||||
XM_011513772.1 | Intron | XP_011512074.1 | ||||
XM_011513773.1 | Intron | XP_011512075.1 |
LOC101929095 - uncharacterized LOC101929095 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |