Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STRIP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STRIP2 - striatin interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134336.1 | Intron | NP_001127808.1 | ||||
NM_020704.2 | Intron | NP_065755.1 | ||||
XM_006716069.3 | Intron | XP_006716132.1 | ||||
XM_011516432.2 | Intron | XP_011514734.1 | ||||
XM_011516435.1 | Intron | XP_011514737.1 | ||||
XM_017012470.1 | Intron | XP_016867959.1 | ||||
XM_017012471.1 | Intron | XP_016867960.1 | ||||
XM_017012472.1 | Intron | XP_016867961.1 |