Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114213 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CALD1 - caldesmon 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004342.6 | Intron | NP_004333.1 | ||||
NM_033138.3 | Intron | NP_149129.2 | ||||
NM_033139.3 | Intron | NP_149130.1 | ||||
NM_033140.3 | Intron | NP_149131.1 | ||||
NM_033157.3 | Intron | NP_149347.2 | ||||
XM_017012650.1 | Intron | XP_016868139.1 | ||||
XM_017012651.1 | Intron | XP_016868140.1 | ||||
XM_017012652.1 | Intron | XP_016868141.1 | ||||
XM_017012653.1 | Intron | XP_016868142.1 | ||||
XM_017012654.1 | Intron | XP_016868143.1 |
Set Membership: |
HapMap |