Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606868 MIM: 274180 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIPK2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HIPK2 - homeodomain interacting protein kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBXAS1 - thromboxane A synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001061.4 | Intron | NP_001052.2 | ||||
NM_001130966.2 | Intron | NP_001124438.1 | ||||
NM_001166253.1 | Intron | NP_001159725.1 | ||||
NM_001166254.1 | Intron | NP_001159726.1 | ||||
NM_001314028.1 | Intron | NP_001300957.1 | ||||
NM_030984.3 | Intron | NP_112246.2 | ||||
XM_011516544.2 | Intron | XP_011514846.1 | ||||
XM_017012569.1 | Intron | XP_016868058.1 | ||||
XM_017012570.1 | Intron | XP_016868059.1 | ||||
XM_017012571.1 | Intron | XP_016868060.1 | ||||
XM_017012572.1 | Intron | XP_016868061.1 |
Set Membership: |
HapMap |