Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606350 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
APTX PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
APTX - aprataxin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195248.1 | 4135 | Intron | NP_001182177.1 | |||
NM_001195249.1 | 4135 | Intron | NP_001182178.1 | |||
NM_001195250.1 | 4135 | Intron | NP_001182179.1 | |||
NM_001195251.1 | 4135 | Intron | NP_001182180.1 | |||
NM_001195252.1 | 4135 | Intron | NP_001182181.1 | |||
NM_001195254.1 | 4135 | Intron | NP_001182183.1 | |||
NM_175069.2 | 4135 | Intron | NP_778239.1 | |||
NM_175073.2 | 4135 | Intron | NP_778243.1 | |||
XM_006716791.3 | 4135 | Intron | XP_006716854.1 | |||
XM_006716792.2 | 4135 | Intron | XP_006716855.1 | |||
XM_011517938.1 | 4135 | Intron | XP_011516240.1 | |||
XM_011517939.2 | 4135 | Intron | XP_011516241.1 | |||
XM_017014831.1 | 4135 | UTR 3 | XP_016870320.1 | |||
XM_017014832.1 | 4135 | Intron | XP_016870321.1 | |||
XM_017014833.1 | 4135 | Intron | XP_016870322.1 | |||
XM_017014834.1 | 4135 | Intron | XP_016870323.1 | |||
XM_017014835.1 | 4135 | Intron | XP_016870324.1 | |||
XM_017014836.1 | 4135 | Intron | XP_016870325.1 | |||
XM_017014837.1 | 4135 | Intron | XP_016870326.1 | |||
XM_017014838.1 | 4135 | Intron | XP_016870327.1 |
Set Membership: |
HapMap |