Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614631 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISPD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ISPD - isoprenoid synthase domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101417.3 | Intron | NP_001094887.1 | ||||
NM_001101426.3 | Intron | NP_001094896.1 | ||||
XM_006715770.3 | Intron | XP_006715833.1 | ||||
XM_011515499.2 | Intron | XP_011513801.1 | ||||
XM_011515500.2 | Intron | XP_011513802.1 | ||||
XM_011515501.2 | Intron | XP_011513803.1 | ||||
XM_011515508.2 | Intron | XP_011513810.1 | ||||
XM_011515509.2 | Intron | XP_011513811.1 | ||||
XM_017012575.1 | Intron | XP_016868064.1 | ||||
XM_017012576.1 | Intron | XP_016868065.1 | ||||
XM_017012577.1 | Intron | XP_016868066.1 | ||||
XM_017012578.1 | Intron | XP_016868067.1 |
Set Membership: |
HapMap |