Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610914 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENOX1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ENOX1 - ecto-NOX disulfide-thiol exchanger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127615.1 | Intron | NP_001121087.1 | ||||
NM_001242863.1 | Intron | NP_001229792.1 | ||||
NM_017993.3 | Intron | NP_060463.2 | ||||
XM_005266439.3 | Intron | XP_005266496.1 | ||||
XM_011535126.2 | Intron | XP_011533428.1 | ||||
XM_011535127.2 | Intron | XP_011533429.1 | ||||
XM_011535128.2 | Intron | XP_011533430.1 | ||||
XM_011535132.2 | Intron | XP_011533434.1 | ||||
XM_017020637.1 | Intron | XP_016876126.1 | ||||
XM_017020638.1 | Intron | XP_016876127.1 | ||||
XM_017020639.1 | Intron | XP_016876128.1 | ||||
XM_017020640.1 | Intron | XP_016876129.1 | ||||
XM_017020641.1 | Intron | XP_016876130.1 | ||||
XM_017020642.1 | Intron | XP_016876131.1 |
Set Membership: |
HapMap |