Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATTATTTCCAACCCTGAGAATCC[A/G]TAATAGATTTGTTTAAAATGACCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD21 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KCTD21 - potassium channel tetramerization domain containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029859.2 | 1699 | UTR 3 | NP_001025030.1 | |||
XM_005273925.3 | 1699 | UTR 3 | XP_005273982.1 | |||
XM_005273928.1 | 1699 | UTR 3 | XP_005273985.1 | |||
XM_006718516.3 | 1699 | UTR 3 | XP_006718579.1 | |||
XM_006718517.2 | 1699 | UTR 3 | XP_006718580.1 | |||
XM_006718518.3 | 1699 | UTR 3 | XP_006718581.1 | |||
XM_011544955.2 | 1699 | UTR 3 | XP_011543257.1 |
KCTD21-AS1 - KCTD21 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |