Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612436 | ||||||||||||||||||||
Literature Links: |
SLCO4A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLCO4A1 - solute carrier organic anion transporter family member 4A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016354.3 | Intron | NP_057438.3 | ||||
XM_005260203.2 | Intron | XP_005260260.1 | ||||
XM_011528792.2 | Intron | XP_011527094.1 | ||||
XM_017027826.1 | Intron | XP_016883315.1 | ||||
XM_017027827.1 | Intron | XP_016883316.1 |