Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LONRF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LONRF1 - LON peptidase N-terminal domain and ring finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005273685.3 | Intron | XP_005273742.1 | ||||
XM_011544694.2 | Intron | XP_011542996.1 |
MIR3926-1 - microRNA 3926-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3926-2 - microRNA 3926-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5692A2 - microRNA 5692a-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |