Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR2H1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OR2H1 - olfactory receptor family 2 subfamily H member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318014.1 | Intron | NP_001304943.1 | ||||
NM_001318022.1 | Intron | NP_001304951.1 | ||||
NM_030883.4 | Intron | NP_112145.1 | ||||
XM_011514470.2 | Intron | XP_011512772.1 | ||||
XM_011514472.2 | Intron | XP_011512774.1 | ||||
XM_011514473.2 | Intron | XP_011512775.1 | ||||
XM_011514479.2 | Intron | XP_011512781.1 | ||||
XM_011514483.2 | Intron | XP_011512785.1 | ||||
XM_011514484.2 | Intron | XP_011512786.1 | ||||
XM_017010737.1 | Intron | XP_016866226.1 | ||||
XM_017010738.1 | Intron | XP_016866227.1 | ||||
XM_017010739.1 | Intron | XP_016866228.1 | ||||
XM_017010740.1 | Intron | XP_016866229.1 |
OR5V1 - olfactory receptor family 5 subfamily V member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |